Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36265669-36265894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL1
Entrez Gene ID
8542
Gene Name
apolipoprotein L1
Gene Aliases
APO-L, APOL, APOL-I, FSGS4
UniGene
Hs.114309
Gene Chromosome Location
Chr.22: 36253071-36267531 on Build GRCh38
Transcripts
NM_001136540.1 NM_001136541.1 NM_003661.3 NM_145343.2 XM_005261796.3 XM_011530478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6503413 c.1008G>A p.R336R Chr.22: 36265796-36265796 on Build GRCh38
3553933 c.985G>A p.E329K Chr.22: 36265773-36265773 on Build GRCh38
3553934 c.1002G>A p.V334V Chr.22: 36265790-36265790 on Build GRCh38
4987852 c.1072A>G p.S358G Chr.22: 36265860-36265860 on Build GRCh38
4103810 c.894C>T p.D298D Chr.22: 36265682-36265682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148496792 Chr.22: 36265727-36265727 on Build GRCh38
rs750486483 Chr.22: 36265847-36265847 on Build GRCh38
rs553515630 Chr.22: 36265843-36265843 on Build GRCh38
rs141788376 Chr.22: 36265745-36265745 on Build GRCh38
rs16996616 Chr.22: 36265845-36265845 on Build GRCh38
rs760331517 Chr.22: 36265838-36265838 on Build GRCh38
rs768179167 Chr.22: 36265730-36265730 on Build GRCh38
rs369288414 Chr.22: 36265699-36265699 on Build GRCh38
rs747821454 Chr.22: 36265811-36265811 on Build GRCh38
rs747993695 Chr.22: 36265869-36265869 on Build GRCh38
rs776081356 Chr.22: 36265841-36265841 on Build GRCh38
rs963940689 Chr.22: 36265851-36265851 on Build GRCh38
rs781041093 Chr.22: 36265718-36265718 on Build GRCh38
rs139590386 Chr.22: 36265764-36265764 on Build GRCh38
rs757346569 Chr.22: 36265713-36265713 on Build GRCh38
rs774066225 Chr.22: 36265731-36265731 on Build GRCh38
rs373044647 Chr.22: 36265844-36265844 on Build GRCh38
rs780096138 Chr.22: 36265779-36265779 on Build GRCh38
rs139583750 Chr.22: 36265839-36265839 on Build GRCh38
rs771843280 Chr.22: 36265871-36265871 on Build GRCh38
rs778126562 Chr.22: 36265705-36265705 on Build GRCh38
rs200272963 Chr.22: 36265729-36265729 on Build GRCh38
rs772882009 Chr.22: 36265749-36265749 on Build GRCh38
rs370479669 Chr.22: 36265756-36265756 on Build GRCh38
rs764169216 Chr.22: 36265763-36265763 on Build GRCh38
rs779788886 Chr.22: 36265858-36265858 on Build GRCh38
rs969002498 Chr.22: 36265861-36265861 on Build GRCh38
rs758858193 Chr.22: 36265760-36265760 on Build GRCh38
rs887099521 Chr.22: 36265805-36265805 on Build GRCh38
rs761340027 Chr.22: 36265741-36265741 on Build GRCh38
rs150588135 Chr.22: 36265750-36265750 on Build GRCh38
rs747082364 Chr.22: 36265709-36265709 on Build GRCh38
rs563861690 Chr.22: 36265766-36265766 on Build GRCh38
rs751786377 Chr.22: 36265863-36265863 on Build GRCh38
rs373769675 Chr.22: 36265778-36265778 on Build GRCh38
rs765764553 Chr.22: 36265744-36265744 on Build GRCh38
rs373409702 Chr.22: 36265864-36265864 on Build GRCh38
rs1003972112 Chr.22: 36265728-36265728 on Build GRCh38
rs924369031 Chr.22: 36265743-36265743 on Build GRCh38
rs1010739230 Chr.22: 36265748-36265748 on Build GRCh38
rs189859721 Chr.22: 36265726-36265726 on Build GRCh38
rs756230453 Chr.22: 36265857-36265857 on Build GRCh38
rs760094738 Chr.22: 36265746-36265746 on Build GRCh38
rs144391919 Chr.22: 36265768-36265768 on Build GRCh38
rs755351451 Chr.22: 36265692-36265692 on Build GRCh38
rs136177 Chr.22: 36265796-36265796 on Build GRCh38
rs186196175 Chr.22: 36265698-36265698 on Build GRCh38
rs73885319 Chr.22: 36265860-36265860 on Build GRCh38
rs200395416 Chr.22: 36265803-36265803 on Build GRCh38
rs370571630 Chr.22: 36265831-36265831 on Build GRCh38

View Full Product Details