Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110650949-110651220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
UniGene
Hs.508725
Gene Chromosome Location
Chr.13: 110641410-110713527 on Build GRCh38
Transcripts
NM_024537.3 XM_006719953.2 XM_011521116.1 XM_017020741.1 XM_017020742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5159793 c.1011delC p.D338fs*44 Chr.13: 110651077-110651077 on Build GRCh38
4902446 c.1051T>C p.S351P Chr.13: 110651037-110651037 on Build GRCh38
2263258 c.1036C>T p.R346W Chr.13: 110651052-110651052 on Build GRCh38
4669309 c.1005T>C p.F335F Chr.13: 110651083-110651083 on Build GRCh38
4669308 c.1048C>T p.R350C Chr.13: 110651040-110651040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769930280 Chr.13: 110651002-110651002 on Build GRCh38
rs376541901 Chr.13: 110651151-110651151 on Build GRCh38
rs192671914 Chr.13: 110650993-110650993 on Build GRCh38
rs758020251 Chr.13: 110651074-110651074 on Build GRCh38
rs768819022 Chr.13: 110651111-110651112 on Build GRCh38
rs550120861 Chr.13: 110650976-110650976 on Build GRCh38
rs767988744 Chr.13: 110651056-110651056 on Build GRCh38
rs71669698 Chr.13: 110650999-110650999 on Build GRCh38
rs756714802 Chr.13: 110651007-110651007 on Build GRCh38
rs974346491 Chr.13: 110651179-110651179 on Build GRCh38
rs201251622 Chr.13: 110651111-110651111 on Build GRCh38
rs748718611 Chr.13: 110650994-110650994 on Build GRCh38
rs774428887 Chr.13: 110651124-110651124 on Build GRCh38
rs189897575 Chr.13: 110651006-110651006 on Build GRCh38
rs374859 Chr.13: 110651026-110651026 on Build GRCh38
rs775870886 Chr.13: 110651143-110651143 on Build GRCh38
rs577161897 Chr.13: 110651076-110651076 on Build GRCh38
rs1005172422 Chr.13: 110650978-110650978 on Build GRCh38
rs138951398 Chr.13: 110651067-110651067 on Build GRCh38
rs748053250 Chr.13: 110651033-110651033 on Build GRCh38
rs751045597 Chr.13: 110651152-110651152 on Build GRCh38
rs763104802 Chr.13: 110651003-110651003 on Build GRCh38
rs548292341 Chr.13: 110651004-110651004 on Build GRCh38
rs185015428 Chr.13: 110651000-110651000 on Build GRCh38
rs746324498 Chr.13: 110650990-110651012 on Build GRCh38
rs764664892 Chr.13: 110651148-110651148 on Build GRCh38
rs753004657 Chr.13: 110651066-110651066 on Build GRCh38
rs773315723 Chr.13: 110651051-110651051 on Build GRCh38
rs778494985 Chr.13: 110651014-110651014 on Build GRCh38
rs773231493 Chr.13: 110651040-110651040 on Build GRCh38
rs201352308 Chr.13: 110651019-110651019 on Build GRCh38
rs751834647 Chr.13: 110651005-110651005 on Build GRCh38
rs772193623 Chr.13: 110651128-110651128 on Build GRCh38
rs763055454 Chr.13: 110651047-110651047 on Build GRCh38
rs932427630 Chr.13: 110651016-110651016 on Build GRCh38
rs766461148 Chr.13: 110651127-110651127 on Build GRCh38
rs780988999 Chr.13: 110651031-110651031 on Build GRCh38
rs755391872 Chr.13: 110650983-110650983 on Build GRCh38
rs771113947 Chr.13: 110651050-110651050 on Build GRCh38
rs369116566 Chr.13: 110651027-110651027 on Build GRCh38
rs376496407 Chr.13: 110651149-110651149 on Build GRCh38
rs568891064 Chr.13: 110651110-110651110 on Build GRCh38
rs759804272 Chr.13: 110651052-110651052 on Build GRCh38
rs768436798 Chr.13: 110651001-110651001 on Build GRCh38
rs756838592 Chr.13: 110650996-110650996 on Build GRCh38
rs574910161 Chr.13: 110651104-110651104 on Build GRCh38
rs778693201 Chr.13: 110650999-110650999 on Build GRCh38
rs771058752 Chr.13: 110651118-110651118 on Build GRCh38
rs145909940 Chr.13: 110651038-110651038 on Build GRCh38
rs556565233 Chr.13: 110651081-110651081 on Build GRCh38
rs1052359973 Chr.13: 110651048-110651048 on Build GRCh38
rs142857490 Chr.13: 110651077-110651077 on Build GRCh38
rs373398364 Chr.13: 110651075-110651075 on Build GRCh38
rs199844548 Chr.13: 110651144-110651144 on Build GRCh38
rs772521220 Chr.13: 110651018-110651018 on Build GRCh38
rs772172464 Chr.13: 110650998-110650999 on Build GRCh38
rs779882291 Chr.13: 110651021-110651021 on Build GRCh38
rs749912792 Chr.13: 110651068-110651068 on Build GRCh38
rs777731745 Chr.13: 110651085-110651085 on Build GRCh38

View Full Product Details