Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 18877043-18877316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH4A1
Entrez Gene ID
8659
Gene Name
aldehyde dehydrogenase 4 family member A1
Gene Aliases
ALDH4, P5CD, P5CDh
UniGene
Hs.77448
Gene Chromosome Location
Chr.1: 18871430-18902799 on Build GRCh38
Transcripts
NM_001161504.1 NM_001319218.1 NM_003748.3 NM_170726.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1134408 c.1166C>G p.S389C Chr.1: 18877227-18877227 on Build GRCh38
458341 c.1166C>G p.S389C Chr.1: 18877227-18877227 on Build GRCh38
2123843 c.1164C>G p.F388L Chr.1: 18877229-18877229 on Build GRCh38
2123844 c.1164C>G p.F388L Chr.1: 18877229-18877229 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762594300 Chr.1: 18877159-18877159 on Build GRCh38
rs767665046 Chr.1: 18877212-18877213 on Build GRCh38
rs754377425 Chr.1: 18877157-18877157 on Build GRCh38
rs899497437 Chr.1: 18877147-18877147 on Build GRCh38
rs930165447 Chr.1: 18877067-18877067 on Build GRCh38
rs769035060 Chr.1: 18877221-18877221 on Build GRCh38
rs757119976 Chr.1: 18877271-18877271 on Build GRCh38
rs1039450828 Chr.1: 18877136-18877136 on Build GRCh38
rs755890443 Chr.1: 18877261-18877261 on Build GRCh38
rs751050137 Chr.1: 18877238-18877238 on Build GRCh38
rs749181166 Chr.1: 18877193-18877193 on Build GRCh38
rs959424547 Chr.1: 18877145-18877145 on Build GRCh38
rs777725664 Chr.1: 18877188-18877188 on Build GRCh38
rs542189193 Chr.1: 18877284-18877284 on Build GRCh38
rs766004973 Chr.1: 18877236-18877236 on Build GRCh38
rs775523746 Chr.1: 18877228-18877230 on Build GRCh38
rs566165363 Chr.1: 18877122-18877122 on Build GRCh38
rs533522724 Chr.1: 18877183-18877183 on Build GRCh38
rs1034518189 Chr.1: 18877080-18877080 on Build GRCh38
rs759502315 Chr.1: 18877191-18877192 on Build GRCh38
rs377293806 Chr.1: 18877187-18877187 on Build GRCh38
rs200076678 Chr.1: 18877208-18877208 on Build GRCh38
rs760988407 Chr.1: 18877266-18877267 on Build GRCh38
rs574681452 Chr.1: 18877275-18877275 on Build GRCh38
rs777312198 Chr.1: 18877222-18877222 on Build GRCh38
rs912715020 Chr.1: 18877087-18877087 on Build GRCh38
rs754534234 Chr.1: 18877171-18877171 on Build GRCh38
rs201079601 Chr.1: 18877198-18877198 on Build GRCh38
rs746106042 Chr.1: 18877204-18877204 on Build GRCh38
rs753784956 Chr.1: 18877268-18877268 on Build GRCh38
rs41273175 Chr.1: 18877231-18877231 on Build GRCh38
rs747216007 Chr.1: 18877292-18877292 on Build GRCh38
rs537654831 Chr.1: 18877209-18877209 on Build GRCh38
rs943821297 Chr.1: 18877130-18877130 on Build GRCh38
rs371743661 Chr.1: 18877239-18877239 on Build GRCh38
rs377590498 Chr.1: 18877259-18877259 on Build GRCh38
rs370298372 Chr.1: 18877273-18877273 on Build GRCh38
rs982938643 Chr.1: 18877079-18877079 on Build GRCh38
rs781048869 Chr.1: 18877172-18877172 on Build GRCh38
rs777494577 Chr.1: 18877263-18877263 on Build GRCh38
rs767120058 Chr.1: 18877247-18877247 on Build GRCh38
rs765949903 Chr.1: 18877164-18877164 on Build GRCh38
rs994597209 Chr.1: 18877177-18877177 on Build GRCh38
rs751282737 Chr.1: 18877168-18877168 on Build GRCh38
rs752489380 Chr.1: 18877184-18877184 on Build GRCh38

View Full Product Details