Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16206236-16206509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3 XM_017027420.1 XM_017027421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3692459 c.224C>T p.A75V Chr.19: 16206365-16206365 on Build GRCh38
1733514 c.229G>A p.V77M Chr.19: 16206370-16206370 on Build GRCh38
4823795 c.233C>T p.S78L Chr.19: 16206374-16206374 on Build GRCh38
4662223 c.228C>T p.C76C Chr.19: 16206369-16206369 on Build GRCh38
7293004 c.229G>A p.V77M Chr.19: 16206370-16206370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201614039 Chr.19: 16206417-16206417 on Build GRCh38
rs554220207 Chr.19: 16206332-16206332 on Build GRCh38
rs893985552 Chr.19: 16206422-16206422 on Build GRCh38
rs199500241 Chr.19: 16206385-16206385 on Build GRCh38
rs200974592 Chr.19: 16206441-16206441 on Build GRCh38
rs749198263 Chr.19: 16206292-16206292 on Build GRCh38
rs773282078 Chr.19: 16206316-16206316 on Build GRCh38
rs988117238 Chr.19: 16206374-16206374 on Build GRCh38
rs759974900 Chr.19: 16206331-16206331 on Build GRCh38
rs992476982 Chr.19: 16206392-16206392 on Build GRCh38
rs557270397 Chr.19: 16206366-16206366 on Build GRCh38
rs990643919 Chr.19: 16206274-16206274 on Build GRCh38
rs758149431 Chr.19: 16206436-16206436 on Build GRCh38
rs138900879 Chr.19: 16206375-16206375 on Build GRCh38
rs770958269 Chr.19: 16206329-16206329 on Build GRCh38
rs777123384 Chr.19: 16206330-16206330 on Build GRCh38
rs185628212 Chr.19: 16206405-16206405 on Build GRCh38
rs958351649 Chr.19: 16206388-16206388 on Build GRCh38
rs755026389 Chr.19: 16206369-16206369 on Build GRCh38
rs760830967 Chr.19: 16206322-16206322 on Build GRCh38
rs572975680 Chr.19: 16206424-16206424 on Build GRCh38
rs778991416 Chr.19: 16206370-16206370 on Build GRCh38
rs768576261 Chr.19: 16206296-16206296 on Build GRCh38
rs376660303 Chr.19: 16206338-16206338 on Build GRCh38
rs749998518 Chr.19: 16206364-16206364 on Build GRCh38
rs1019694021 Chr.19: 16206281-16206281 on Build GRCh38
rs777583600 Chr.19: 16206437-16206437 on Build GRCh38
rs747130464 Chr.19: 16206393-16206393 on Build GRCh38
rs561896594 Chr.19: 16206477-16206477 on Build GRCh38
rs369179822 Chr.19: 16206430-16206430 on Build GRCh38
rs150267214 Chr.19: 16206363-16206363 on Build GRCh38
rs200386274 Chr.19: 16206372-16206372 on Build GRCh38
rs753840973 Chr.19: 16206368-16206368 on Build GRCh38
rs763199908 Chr.19: 16206357-16206357 on Build GRCh38
rs760347887 Chr.19: 16206431-16206431 on Build GRCh38
rs377648467 Chr.19: 16206421-16206421 on Build GRCh38
rs374005098 Chr.19: 16206418-16206418 on Build GRCh38
rs771009421 Chr.19: 16206395-16206395 on Build GRCh38
rs202058849 Chr.19: 16206313-16206313 on Build GRCh38
rs772377095 Chr.19: 16206315-16206315 on Build GRCh38
rs778795131 Chr.19: 16206302-16206302 on Build GRCh38
rs766118232 Chr.19: 16206434-16206434 on Build GRCh38

View Full Product Details