Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11300410-11300683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMELX
Entrez Gene ID
265
Gene Name
amelogenin, X-linked
Gene Aliases
AI1E, AIH1, ALGN, AMG, AMGL, AMGX
UniGene
Hs.654436
Gene Chromosome Location
Chr.X: 11293413-11300761 on Build GRCh38
Transcripts
NM_001142.2 NM_182680.1 NM_182681.1 NM_001287242.1 NM_006125.2 NM_013427.2 NR_109776.1

Gene Symbol
ARHGAP6
Entrez Gene ID
395
Gene Name
Rho GTPase activating protein 6
Gene Aliases
RHOGAP6, RHOGAPX-1
UniGene
Hs.435291
Gene Chromosome Location
Chr.X: 11137543-11665701 on Build GRCh38
Transcripts
NM_001142.2 NM_182680.1 NM_182681.1 NM_001287242.1 NM_006125.2 NM_013427.2 NR_109776.1

SNP Detail

SNP ID Chromosome Location
rs889097470 Chr.X: 11300490-11300490 on Build GRCh38
rs769206570 Chr.X: 11300585-11300585 on Build GRCh38
rs771336026 Chr.X: 11300558-11300558 on Build GRCh38
rs1025400520 Chr.X: 11300631-11300631 on Build GRCh38
rs746272574 Chr.X: 11300569-11300569 on Build GRCh38
rs781175862 Chr.X: 11300648-11300648 on Build GRCh38
rs935201154 Chr.X: 11300434-11300434 on Build GRCh38
rs191399494 Chr.X: 11300478-11300478 on Build GRCh38
rs779253128 Chr.X: 11300565-11300565 on Build GRCh38
rs1056208285 Chr.X: 11300481-11300482 on Build GRCh38
rs867629707 Chr.X: 11300632-11300632 on Build GRCh38
rs764911951 Chr.X: 11300606-11300606 on Build GRCh38
rs776065625 Chr.X: 11300594-11300594 on Build GRCh38
rs761293454 Chr.X: 11300602-11300602 on Build GRCh38
rs868271285 Chr.X: 11300633-11300633 on Build GRCh38
rs751302355 Chr.X: 11300630-11300630 on Build GRCh38
rs200715488 Chr.X: 11300650-11300650 on Build GRCh38
rs746508249 Chr.X: 11300456-11300456 on Build GRCh38
rs762731577 Chr.X: 11300597-11300597 on Build GRCh38
rs754937993 Chr.X: 11300637-11300637 on Build GRCh38
rs1051120402 Chr.X: 11300485-11300485 on Build GRCh38
rs896269166 Chr.X: 11300492-11300492 on Build GRCh38
rs372488657 Chr.X: 11300626-11300626 on Build GRCh38
rs183940749 Chr.X: 11300583-11300583 on Build GRCh38
rs367656319 Chr.X: 11300449-11300449 on Build GRCh38
rs376830760 Chr.X: 11300570-11300570 on Build GRCh38
rs369247345 Chr.X: 11300651-11300651 on Build GRCh38
rs1006646194 Chr.X: 11300505-11300505 on Build GRCh38
rs750108771 Chr.X: 11300625-11300625 on Build GRCh38
rs369891804 Chr.X: 11300574-11300574 on Build GRCh38

View Full Product Details