Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83120560-83120833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY1
Entrez Gene ID
23033
Gene Name
dopey family member 1
Gene Aliases
DOP1, KIAA1117, dJ202D23.2
UniGene
Hs.520246
Gene Chromosome Location
Chr.6: 83067650-83171351 on Build GRCh38
Transcripts
NM_001199942.1 NM_015018.3 XM_011535619.2 XM_017010559.1 XM_017010560.1 XM_017010561.1 XM_017010562.1 XM_017010563.1 XM_017010564.1 XM_017010565.1 XM_017010566.1 XM_017010567.1 XM_017010568.1 XM_017010569.1 XM_017010570.1 XM_017010571.1 XM_017010572.1 XM_017010573.1 XM_017010574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3411308 c.1036C>T p.R346C Chr.6: 83120755-83120755 on Build GRCh38
5208476 c.1024C>G p.L342V Chr.6: 83120743-83120743 on Build GRCh38
2151670 c.1063C>T p.R355C Chr.6: 83120755-83120755 on Build GRCh38
3875763 c.1046A>G p.Q349R Chr.6: 83120738-83120738 on Build GRCh38
451835 c.1051C>G p.L351V Chr.6: 83120743-83120743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751909773 Chr.6: 83120739-83120739 on Build GRCh38
rs1046564971 Chr.6: 83120652-83120652 on Build GRCh38
rs771145551 Chr.6: 83120797-83120797 on Build GRCh38
rs764940355 Chr.6: 83120703-83120703 on Build GRCh38
rs371712101 Chr.6: 83120758-83120758 on Build GRCh38
rs781450387 Chr.6: 83120633-83120633 on Build GRCh38
rs760563023 Chr.6: 83120666-83120666 on Build GRCh38
rs756258407 Chr.6: 83120786-83120786 on Build GRCh38
rs745741817 Chr.6: 83120658-83120658 on Build GRCh38
rs745349190 Chr.6: 83120695-83120695 on Build GRCh38
rs531365171 Chr.6: 83120717-83120717 on Build GRCh38
rs1055774755 Chr.6: 83120774-83120774 on Build GRCh38
rs775306770 Chr.6: 83120660-83120660 on Build GRCh38
rs954070452 Chr.6: 83120615-83120620 on Build GRCh38
rs757185380 Chr.6: 83120600-83120600 on Build GRCh38
rs761303438 Chr.6: 83120694-83120694 on Build GRCh38
rs750016824 Chr.6: 83120712-83120712 on Build GRCh38
rs146788469 Chr.6: 83120775-83120775 on Build GRCh38
rs748169584 Chr.6: 83120639-83120639 on Build GRCh38
rs532242271 Chr.6: 83120588-83120588 on Build GRCh38
rs368164785 Chr.6: 83120643-83120643 on Build GRCh38
rs552343766 Chr.6: 83120593-83120593 on Build GRCh38
rs776134113 Chr.6: 83120688-83120688 on Build GRCh38
rs906653819 Chr.6: 83120680-83120680 on Build GRCh38
rs779719636 Chr.6: 83120803-83120803 on Build GRCh38
rs768515488 Chr.6: 83120686-83120686 on Build GRCh38
rs781740683 Chr.6: 83120771-83120771 on Build GRCh38
rs778180229 Chr.6: 83120787-83120787 on Build GRCh38
rs908282942 Chr.6: 83120597-83120597 on Build GRCh38
rs749494633 Chr.6: 83120792-83120792 on Build GRCh38
rs192342724 Chr.6: 83120657-83120657 on Build GRCh38
rs771741933 Chr.6: 83120659-83120659 on Build GRCh38
rs113362312 Chr.6: 83120807-83120807 on Build GRCh38
rs147534184 Chr.6: 83120621-83120621 on Build GRCh38
rs763420601 Chr.6: 83120715-83120715 on Build GRCh38
rs1011347998 Chr.6: 83120589-83120589 on Build GRCh38

View Full Product Details