The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003922.1 | 3 - 4 | 1664 | 79 | |
GenBank mRNA | AY008413.1 | - | 3 - 4 | 1664 | 79 |
EU921431.1 | - | 4 - 5 | 376 | 79 |