The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000968.3 | NP_000959.2 | 6 - 7 | 740 | 113 |
GenBank mRNA | AB208977.1 | - | 6 - 7 | 717 | 113 |
AK125772.1 | - | 7 - 8 | 1171 | 113 | |
AK130784.1 | - | 6 - 7 | 731 | 113 | |
AK223057.1 | - | 6 - 7 | 731 | 113 | |
AK291117.1 | - | 6 - 7 | 749 | 113 | |
AK291859.1 | - | 6 - 7 | 731 | 113 | |
AK297484.1 | - | 2 - 2 | 551 | 113 | |
BC001365.2 | - | 6 - 7 | 721 | 113 | |
BC005817.2 | - | 1 - 1 | 708 | 113 | |
BC007748.2 | - | 1 - 1 | 762 | 113 | |
BC007996.1 | - | 1 - 1 | 714 | 113 | |
BC009888.2 | - | 6 - 7 | 708 | 113 | |
BC010151.2 | - | 6 - 7 | 713 | 113 | |
BC014653.1 | - | 6 - 7 | 709 | 113 | |
BC018840.2 | - | 6 - 7 | 721 | 113 | |
BC040610.1 | - | 1 - 1 | 716 | 113 | |
BC066925.1 | - | 6 - 7 | 731 | 113 | |
BC095427.1 | - | 6 - 7 | 724 | 113 | |
D23660.1 | - | 6 - 7 | 731 | 113 | |
L20868.2 | - | 6 - 7 | 687 | 113 |