Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANKRD30BP2
- Entrez Gene ID
-
149992
- Gene Name
- ankyrin repeat domain 30B pseudogene 2
- Gene Aliases
-
C21orf99, CT85, CTSP-1, CTSP1
- Chromosome Location
-
Chr.21: 13038166 - 13118251 on Build GRCh38
- Gene Symbol
- ANKRD30BL
- Entrez Gene ID
-
554226
- Gene Name
- ankyrin repeat domain 30B-like
- Gene Aliases
-
ANKRD30BP3, NCRNA00164
- Chromosome Location
-
Chr.2: 132147591 - 132257969 on Build GRCh38
View Full Product Details