The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006191.2 | NP_006182.2 | 13 - 13 | 1862 | 112 |
GenBank mRNA | BC069786.1 | - | 13 - 13 | 1515 | 112 |
U59435.1 | - | 13 - 13 | 1540 | 113 |