The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278531.1 | NP_001265460.1 | 13 - 14 | 1352 | 74 |
NM_001278532.1 | NP_001265461.1 | 12 - 13 | 1183 | 74 | |
NM_001278533.1 | NP_001265462.1 | 13 - 14 | 1352 | 74 | |
NM_018341.2 | NP_060811.1 | 13 - 14 | 1352 | 74 | |
XM_011535938.2 | XP_011534240.1 | 12 - 13 | 1322 | 74 | |
XM_011535939.2 | XP_011534241.1 | 13 - 14 | 2442 | 74 | |
XM_011535940.2 | XP_011534242.1 | 13 - 14 | 2251 | 74 | |
XM_017011027.1 | XP_016866516.1 | 13 - 14 | 2192 | 74 | |
XM_017011028.1 | XP_016866517.1 | 13 - 14 | 1517 | 74 | |
XM_017011029.1 | XP_016866518.1 | 13 - 14 | 1277 | 74 | |
XM_017011030.1 | XP_016866519.1 | 12 - 13 | 1101 | 74 | |
XM_017011031.1 | XP_016866520.1 | 12 - 13 | 1166 | 74 | |
XM_017011032.1 | XP_016866521.1 | 13 - 14 | 2437 | 74 | |
XM_017011033.1 | XP_016866522.1 | 13 - 14 | 2246 | 74 | |
GenBank mRNA | AK001369.1 | - | 12 - 13 | 1213 | 74 |
AK002014.1 | - | 13 - 14 | 1349 | 74 | |
AK021520.1 | - | 3 - 4 | 1416 | 74 | |
AK294090.1 | - | 12 - 13 | 1180 | 74 | |
BC044812.1 | - | 13 - 14 | 1336 | 74 |