Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080423.3 NP_001073892.3 15 - 16 1819 89
XM_011534141.2 XP_011532443.1 15 - 16 1901 89
XM_011534142.2 XP_011532444.1 15 - 16 1813 89
XM_011534143.2 XP_011532445.1 14 - 15 1775 89
XM_011534145.2 XP_011532447.1 9 - 10 1547 89
XM_011534146.2 XP_011532448.1 14 - 15 2380 89
GenBank mRNA AB051506.2 - 15 - 16 1805 89
AK024507.1 - 2 - 3 159 89

View Full Product Details