Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080423.3 NP_001073892.3 17 - 18 2114 85
XM_011534141.2 XP_011532443.1 17 - 18 2196 85
XM_011534142.2 XP_011532444.1 17 - 18 2108 85
XM_011534143.2 XP_011532445.1 16 - 17 2070 85
XM_011534145.2 XP_011532447.1 11 - 12 1842 85
XM_011534146.2 XP_011532448.1 16 - 17 2675 85
GenBank mRNA AB051506.2 - 17 - 18 2100 85
AK024507.1 - 4 - 5 454 85

View Full Product Details