Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080423.3 NP_001073892.3 5 - 6 524 60
XM_011534141.2 XP_011532443.1 5 - 6 606 60
XM_011534142.2 XP_011532444.1 5 - 6 518 60
XM_011534143.2 XP_011532445.1 5 - 6 606 60
GenBank mRNA AB051506.2 - 5 - 6 510 60

View Full Product Details