The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284260.1 | NP_001271189.1 | 4 - 5 | 1377 | 81 |
NM_001284262.1 | NP_001271191.1 | 4 - 5 | 814 | 81 | |
NM_198285.2 | NP_938026.2 | 4 - 5 | 1313 | 81 | |
XM_005249990.4 | XP_005250047.1 | 4 - 5 | 1310 | 81 | |
XM_011516144.2 | XP_011514446.1 | 4 - 5 | 1374 | 81 | |
XM_011516145.2 | XP_011514447.1 | 4 - 5 | 1374 | 81 | |
XM_011516146.2 | XP_011514448.1 | 4 - 5 | 1310 | 81 | |
XM_011516148.1 | XP_011514450.1 | 4 - 5 | 862 | 81 | |
XM_011516150.1 | XP_011514452.1 | 5 - 6 | 1066 | 81 | |
GenBank mRNA | AK296049.1 | - | 4 - 5 | 970 | 81 |
BC107125.2 | - | 3 - 4 | 710 | 81 |