The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323422.1 | NP_001310351.1 | 5 - 6 | 772 | 129 |
NM_001323423.1 | NP_001310352.1 | 7 - 8 | 917 | 129 | |
NM_004902.3 | NP_004893.1 | 5 - 6 | 772 | 129 | |
NM_184234.2 | NP_909122.1 | 5 - 6 | 772 | 129 | |
NR_040722.1 | 6 - 7 | 845 | 129 | ||
NR_040723.1 | 6 - 7 | 844 | 129 | ||
NR_040724.1 | 6 - 7 | 845 | 129 | ||
XM_006723890.3 | XP_006723953.1 | 5 - 6 | 2352 | 129 | |
XM_006723891.3 | XP_006723954.1 | 6 - 7 | 1375 | 129 | |
XM_011529111.2 | XP_011527413.1 | 6 - 7 | 2424 | 129 | |
XM_017028139.1 | XP_016883628.1 | 7 - 8 | 1741 | 129 | |
XM_017028141.1 | XP_016883630.1 | 5 - 6 | 2352 | 129 | |
XM_017028142.1 | XP_016883631.1 | 6 - 7 | 3248 | 129 | |
XM_017028143.1 | XP_016883632.1 | 6 - 7 | 2424 | 129 | |
XM_017028144.1 | XP_016883633.1 | 7 - 8 | 917 | 129 | |
XM_017028146.1 | XP_016883635.1 | 6 - 7 | 845 | 129 | |
XM_017028147.1 | XP_016883636.1 | 6 - 7 | 768 | 129 | |
GenBank mRNA | AK126158.1 | - | 2 - 3 | 931 | 129 |
AK303910.1 | - | 5 - 6 | 644 | 129 | |
AL833168.1 | - | 5 - 6 | 658 | 129 | |
BC107886.1 | - | 6 - 7 | 554 | 129 | |
BC141835.1 | - | 5 - 6 | 622 | 129 | |
BC158172.1 | - | 5 - 6 | 622 | 129 | |
BX537770.1 | - | 6 - 7 | 764 | 129 | |
BX640714.1 | - | 6 - 7 | 757 | 129 | |
BX640812.1 | - | 6 - 7 | 754 | 129 | |
CR749443.1 | - | 5 - 6 | 2439 | 129 | |
L10910.1 | - | 5 - 6 | 512 | 129 | |
L10911.1 | - | 4 - 5 | 387 | 129 |