The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093725.1 | NP_001087194.1 | 1 - 2 | 455 | 165 |
GenBank mRNA | AY950677.1 | - | 1 - 2 | 455 | 165 |