Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RAET1K
- Entrez Gene ID
-
646024
- Gene Name
- retinoic acid early transcript 1K pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 149998019 - 150005144 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024045.1 |
|
1 - 2 |
72 |
97 |
GenBank mRNA |
AF425244.1 |
- |
1 - 2 |
72 |
97 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI017460.1 |
- |
1 - 2 |
54 |
|
Chr. 6 - 150001259 - 150005126 [-] on Build GRCh38 |
97 |
GenBankmRNA |
AA884304.1 |
- |
1 - 2 |
|
|
Chr. 6 - 150001227 - 150005126 [-] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
UL16 binding protein 1 |
ULBP1 |
N2DL-1, NKG2DL1, RAET1I |
-27.1kb
|
Anti-sense |
Chr. - 149963974 - 149973715 on Build GRCh38 |
Downstream |
retinoic acid early transcript 1L |
RAET1L |
ULBP6 |
13.2kb
|
Sense |
Chr. - 150018334 - 150025596 on Build GRCh38 |
View Full Product Details