Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRB4
Entrez Gene ID
5545
Gene Name
proline rich protein BstNI subfamily 4
Gene Aliases
Po
Chromosome Location
Chr.12: 11307081 - 11310435 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261399.2 NP_001248328.1 4 - 5 587 93
NM_002723.5 NP_002714.2 3 - 4 794 93
GenBank mRNA BC035362.1 - 3 - 4 843 93
BC130386.1 - 3 - 4 842 93

View Full Product Details