The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012249.3 | NP_036381.2 | 5 - 5 | 2124 | 113 |
XM_005264229.2 | XP_005264286.1 | 3 - 3 | 1826 | 113 | |
XM_011532726.2 | XP_011531028.1 | 6 - 6 | 2417 | 113 | |
XM_011532728.2 | XP_011531030.1 | 4 - 4 | 1690 | 113 | |
XM_017003716.1 | XP_016859205.1 | 6 - 6 | 2574 | 113 | |
GenBank mRNA | EU794657.1 | - | 5 - 5 | 2124 | 113 |