Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IGKV3-20
- Entrez Gene ID
-
28912
- Gene Name
- immunoglobulin kappa variable 3-20
- Gene Aliases
-
13K18, A27, IGKV320
- Chromosome Location
-
Chr.2: 89142574 - 89143108 on Build GRCh38
- Gene Symbol
- IGHV3-23
- Entrez Gene ID
-
28442
- Gene Name
- immunoglobulin heavy variable 3-23
- Gene Aliases
-
DP47, IGHV323, V3-23, VH26
- Chromosome Location
-
Chr.14: 106268606 - 106269061 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF231396.1 |
- |
1 - 1 |
252 |
149 |
X14584.1 |
- |
2 - 2 |
333 |
149 |
- Gene Symbol
- ZCWPW2
- Entrez Gene ID
-
152098
- Gene Name
- zinc finger CW-type and PWWP domain containing 2
- Gene Aliases
-
ZCW2
- Chromosome Location
-
Chr.3: 28349149 - 28526366 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB087877.1 |
- |
1 - 1 |
266 |
149 |
View Full Product Details