Product Details

Assay Reference Genome
Location

Chr.8:38412482 on build GRCh38
Cytoband
8p11.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2260
Gene Symbol
FGFR1
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Location
Chr.8:38411138-38468834 on build GRCh38
Assay Gene Location
Within Exon 23
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FGFR1 NM_001174063.1 18 4551 NP_001167534.1
NM_001174064.1 19 4619 NP_001167535.1
NM_001174065.1 18 3871 NP_001167536.1
NM_001174066.1 17 3610 NP_001167537.1
NM_001174067.1 19 4031 NP_001167538.1
NM_015850.3 18 4551 NP_056934.2
NM_023105.2 17 4290 NP_075593.1
NM_023106.2 17 4284 NP_075594.1
NM_023110.2 18 4557 NP_075598.2
XM_006716303.2 18 3149 XP_006716366.1
XM_006716304.1 18 2665 XP_006716367.1
XM_006716306.2 18 3143 XP_006716369.1
XM_006716307.1 18 2659 XP_006716370.1
XM_006716309.3 18 3119 XP_006716372.1
XM_006716310.2 17 2882 XP_006716373.1
XM_006716311.1 17 2280 XP_006716374.1
XM_006716312.1 17 2350 XP_006716375.1
XM_006716313.2 17 2876 XP_006716376.1
XM_006716314.1 17 2354 XP_006716377.1
XM_011544443.2 18 2654 XP_011542745.1
XM_011544444.1 18 2643 XP_011542746.1
XM_011544445.2 18 2656 XP_011542747.1
XM_011544446.2 18 3858 XP_011542748.1
XM_011544447.2 18 3852 XP_011542749.1
XM_011544448.1 17 2253 XP_011542750.1
XM_011544449.1 17 2247 XP_011542751.1
XM_011544450.2 17 3456 XP_011542752.1
XM_011544451.1 16 2116 XP_011542753.1
XM_017013219.1 18 2644 XP_016868708.1
XM_017013220.1 18 3846 XP_016868709.1
XM_017013221.1 19 2858 XP_016868710.1
XM_017013222.1 18 3143 XP_016868711.1
XM_017013223.1 18 2653 XP_016868712.1
XM_017013224.1 18 4346 XP_016868713.1
XM_017013225.1 18 3829 XP_016868714.1
XM_017013226.1 17 3456 XP_016868715.1
XM_017013227.1 17 3450 XP_016868716.1
XM_017013228.1 17 4079 XP_016868717.1
XM_017013229.1 12 2829 XP_016868718.1
XM_017013230.1 16 4644 XP_016868719.1
CX757985.1 1 409
FJ809917.1 18 4566

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951128 Chr8:38411783 - 38418282 on Build GRCh38 Deletion FGFR1

View Full Product Details