The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024241.1 | 8 - 8 | 1649 | 160 | |
GenBank mRNA | BC043342.2 | - | 7 - 7 | 1373 | 160 |