The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304359.1 | NP_001291288.1 | - | 1755 | 86 |
GenBank mRNA | AF015521.1 | - | 14 - 15 | 1755 | 86 |
AF043909.1 | - | 14 - 15 | 1746 | 86 | |
AJ298317.1 | - | 14 - 15 | 1746 | 86 |