Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTAR1
Entrez Gene ID
375743
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 69709522 - 69760050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099666.1 NP_001093136.1 2 - 3 271 89
XM_005251976.4 XP_005252033.1 2 - 3 361 89
XM_005251977.4 XP_005252034.1 2 - 3 335 89
XM_005251980.4 XP_005252037.1 2 - 3 321 89
XM_005251981.4 XP_005252038.1 2 - 3 321 89
XM_011518640.1 XP_011516942.1 2 - 3 355 89
GenBank mRNA BC053622.1 - 2 - 3 253 89

View Full Product Details