The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077706.2 | NP_001071174.1 | 9 - 10 | 1400 | 80 |
NM_001195037.2 | NP_001181966.1 | 8 - 9 | 1260 | 80 | |
XM_006715472.3 | XP_006715535.1 | 9 - 10 | 1393 | 80 | |
XM_011535795.2 | XP_011534097.1 | 8 - 9 | 1264 | 80 | |
XM_011535797.2 | XP_011534099.1 | 6 - 7 | 976 | 80 | |
XM_017010828.1 | XP_016866317.1 | 9 - 10 | 1366 | 80 | |
XM_017010829.1 | XP_016866318.1 | 9 - 10 | 1393 | 80 | |
XM_017010830.1 | XP_016866319.1 | 9 - 10 | 1393 | 80 | |
GenBank mRNA | AK304425.1 | - | 6 - 7 | 958 | 80 |
BC146884.1 | - | 7 - 8 | 1107 | 80 | |
DQ904331.1 | - | 7 - 8 | 1054 | 80 |