Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN9
Entrez Gene ID
392399
Gene Name
lipocalin 9
Gene Aliases
9230102I19Rik, HEL129
Chromosome Location
Chr.9: 135663322 - 135670731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001676.1 NP_001001676.1 2 - 3 232 66
XM_017014712.1 XP_016870201.1 2 - 3 232 66
GenBank mRNA AY301270.1 - 2 - 3 232 66
EU794679.1 - 2 - 3 232 66

View Full Product Details