The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138690.2 | NP_619635.1 | 2 - 3 | 1037 | 113 |
GenBank mRNA | AY507106.1 | - | 2 - 3 | 1037 | 113 |
AY507107.1 | - | 2 - 3 | 1037 | 113 |