Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXND1
Entrez Gene ID
23129
Gene Name
plexin D1
Gene Aliases
PLEXD1
Chromosome Location
Chr.3: 129555213 - 129606739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015103.2 NP_055918.2 8 - 9 2342 108
XM_011512588.1 XP_011510890.1 8 - 9 2257 108
XM_011512590.2 XP_011510892.1 8 - 9 2257 108
GenBank mRNA AB014520.2 - 8 - 9 2421 108
AY116661.1 - 8 - 9 2422 108
BC150280.1 - 8 - 9 2421 108

View Full Product Details