The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015103.2 | NP_055918.2 | 9 - 10 | 2448 | 59 |
XM_011512588.1 | XP_011510890.1 | 9 - 10 | 2363 | 59 | |
XM_011512589.1 | XP_011510891.1 | 6 - 7 | 1973 | 59 | |
XM_011512590.2 | XP_011510892.1 | 9 - 10 | 2363 | 59 | |
GenBank mRNA | AB014520.2 | - | 9 - 10 | 2527 | 59 |
AY116661.1 | - | 9 - 10 | 2528 | 59 | |
BC150280.1 | - | 9 - 10 | 2527 | 59 |