Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRAP73
Entrez Gene ID
49856
Gene Name
WD repeat containing, antisense to TP73
Gene Aliases
WDR8
Chromosome Location
Chr.1: 3630767 - 3650107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017818.3 NP_060288.3 8 - 9 921 61
XM_005244754.1 XP_005244811.1 7 - 8 781 61
XM_017001387.1 XP_016856876.1 8 - 9 916 61
GenBank mRNA AB034912.1 - 8 - 9 872 61
AK000437.1 - 8 - 9 887 61
AK225525.1 - 8 - 9 887 61
BC086311.1 - 8 - 9 894 61
EF494669.1 - 8 - 9 793 61

View Full Product Details