Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL9
Entrez Gene ID
51108
Gene Name
methyltransferase like 9
Gene Aliases
CGI-81, DREV, DREV1, PAP1
Chromosome Location
Chr.16: 21597221 - 21657473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077180.2 NP_001070648.1 2 - 3 616 135
NM_001288659.1 NP_001275588.1 2 - 3 273 135
NM_001288660.1 NP_001275589.1 2 - 3 273 135
NM_016025.4 NP_057109.3 2 - 3 616 135
GenBank mRNA AF151839.1 - 2 - 3 280 135
AJ278577.1 - 2 - 3 534 135
AJ278578.1 - 2 - 3 534 135
AK075022.1 - 2 - 3 502 135
AK075237.1 - 2 - 3 485 135
AK313077.1 - 2 - 3 500 135
BC000195.1 - 2 - 3 501 135
BG183771.1 - 2 - 3 272 135

View Full Product Details