Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC4
Entrez Gene ID
58484
Gene Name
NLR family CARD domain containing 4
Gene Aliases
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF
Chromosome Location
Chr.2: 32224449 - 32265743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199138.1 NP_001186067.1 8 - 9 3046 118
NM_001199139.1 NP_001186068.1 8 - 9 2929 118
NM_001302504.1 NP_001289433.1 7 - 8 1065 118
NM_021209.4 NP_067032.3 8 - 9 3060 118
GenBank mRNA AF376061.1 - 9 - 10 3274 118
AK095467.1 - 8 - 9 3046 118
AK292673.1 - 8 - 9 3031 118
AK314762.1 - 8 - 9 3031 118
AL389934.1 - 5 - 6 1031 118
AY027787.1 - 8 - 9 3061 118
AY027788.1 - 7 - 8 1066 118
AY032589.1 - 7 - 8 2820 118
AY035391.1 - 8 - 9 2929 118
AY358152.1 - 8 - 9 3008 118
BC031555.1 - 8 - 9 3016 118

View Full Product Details