The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253357.1 | NP_001240286.1 | 13 - 14 | 2312 | 75 |
NM_005424.4 | NP_005415.1 | 13 - 14 | 2395 | 75 | |
XM_005271163.1 | XP_005271220.1 | 12 - 13 | 2209 | 75 | |
XM_006710869.1 | XP_006710932.1 | 13 - 14 | 2529 | 75 | |
XM_017002207.1 | XP_016857696.1 | 13 - 14 | 2338 | 75 | |
GenBank mRNA | AF217976.1 | - | 3 - 4 | 479 | 75 |
AK225560.1 | - | 13 - 14 | 2054 | 75 | |
AK296735.1 | - | 8 - 9 | 1371 | 75 | |
AK300399.1 | - | 13 - 14 | 2202 | 75 | |
AK300623.1 | - | 13 - 14 | 2095 | 75 | |
AK307639.1 | - | 6 - 7 | 1063 | 75 | |
BC038239.1 | - | 13 - 14 | 2297 | 75 | |
X60957.1 | - | 13 - 14 | 2254 | 75 |