The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253357.1 | NP_001240286.1 | 16 - 17 | 2832 | 107 |
NM_005424.4 | NP_005415.1 | 16 - 17 | 2915 | 107 | |
XM_005271163.1 | XP_005271220.1 | 15 - 16 | 2729 | 107 | |
XM_006710869.1 | XP_006710932.1 | 16 - 17 | 3049 | 107 | |
GenBank mRNA | AF217976.1 | - | 6 - 7 | 999 | 107 |
AK225560.1 | - | 16 - 17 | 2574 | 107 | |
AK296735.1 | - | 11 - 12 | 1891 | 107 | |
AK300399.1 | - | 16 - 17 | 2722 | 107 | |
AK307639.1 | - | 9 - 10 | 1583 | 107 | |
BC038239.1 | - | 16 - 17 | 2817 | 107 | |
X60957.1 | - | 16 - 17 | 2774 | 107 |