The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253357.1 | NP_001240286.1 | 17 - 18 | 3020 | 71 |
NM_005424.4 | NP_005415.1 | 17 - 18 | 3103 | 71 | |
XM_005271163.1 | XP_005271220.1 | 16 - 17 | 2917 | 71 | |
XM_006710869.1 | XP_006710932.1 | 17 - 18 | 3237 | 71 | |
GenBank mRNA | AF217976.1 | - | 7 - 8 | 1187 | 71 |
AK225560.1 | - | 17 - 18 | 2762 | 71 | |
AK296735.1 | - | 12 - 13 | 2079 | 71 | |
AK300399.1 | - | 17 - 18 | 2910 | 71 | |
AL833389.1 | - | 2 - 3 | 1555 | 71 | |
BC038239.1 | - | 17 - 18 | 3005 | 71 | |
X60957.1 | - | 17 - 18 | 2962 | 71 |