Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
Chromosome Location
Chr.1: 56929210 - 56974383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000066.3 NP_000057.2 1 - 2 281 80
NM_001278544.1 NP_001265473.1 1 - 2 281 80
XM_017002235.1 XP_016857724.1 2 - 3 349 80
GenBank mRNA AK292740.1 - 1 - 2 136 80
AK298425.1 - 1 - 2 198 80
AK313382.1 - 1 - 2 153 80
BC020701.1 - 1 - 2 156 80
BC130575.1 - 1 - 2 114 80
M16973.1 - 1 - 2 116 80

View Full Product Details