Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
Chromosome Location
Chr.1: 56929210 - 56974383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000066.3 NP_000057.2 6 - 7 1058 92
NM_001278543.1 NP_001265472.1 7 - 8 1280 92
NM_001278544.1 NP_001265473.1 7 - 8 1187 92
XM_017002235.1 XP_016857724.1 7 - 8 1126 92
GenBank mRNA AK292740.1 - 6 - 7 913 92
AK298425.1 - 7 - 8 1104 92
AK298444.1 - 7 - 8 1280 92
AK313382.1 - 6 - 7 930 92
AK316209.1 - 7 - 8 1277 92
BC020701.1 - 6 - 7 933 92
BC130575.1 - 6 - 7 891 92
M16973.1 - 6 - 7 893 92
X04393.1 - 5 - 6 728 92

View Full Product Details