Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
Chromosome Location
Chr.1: 56929210 - 56974383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000066.3 NP_000057.2 7 - 8 1295 80
NM_001278543.1 NP_001265472.1 8 - 9 1517 80
NM_001278544.1 NP_001265473.1 8 - 9 1424 80
XM_017002235.1 XP_016857724.1 8 - 9 1363 80
GenBank mRNA AK292740.1 - 7 - 8 1150 80
AK298425.1 - 8 - 9 1341 80
AK298444.1 - 8 - 9 1517 80
AK313382.1 - 7 - 8 1167 80
AK316209.1 - 8 - 9 1514 80
BC020701.1 - 7 - 8 1170 80
BC130575.1 - 7 - 8 1128 80
M16973.1 - 7 - 8 1130 80
X04393.1 - 6 - 7 965 80

View Full Product Details