Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM26
Entrez Gene ID
7726
Gene Name
tripartite motif containing 26
Gene Aliases
AFP, RNF95, ZNF173
Chromosome Location
Chr.6: 30184453 - 30213494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242783.1 NP_001229712.1 7 - 8 1245 112
NM_003449.4 NP_003440.1 8 - 9 1466 112
XM_005249374.2 XP_005249431.1 8 - 9 1262 112
XM_005249375.2 XP_005249432.1 8 - 9 1270 112
XM_005249376.2 XP_005249433.1 8 - 9 1342 112
XM_005249377.2 XP_005249434.1 7 - 8 1102 112
XM_005249378.2 XP_005249435.1 6 - 7 1082 112
XM_006715180.2 XP_006715243.1 7 - 8 1286 112
XM_011514859.1 XP_011513161.1 7 - 8 1031 112
XM_017011263.1 XP_016866752.1 6 - 7 955 112
GenBank mRNA AK096996.1 - 1 - 2 974 112
BC024039.1 - 8 - 9 1320 112
BC032297.1 - 7 - 8 1103 112

View Full Product Details