Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNSD1
Entrez Gene ID
54529
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
NBLA00058, NS3TP1
Chromosome Location
Chr.2: 189661399 - 189670831 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019048.2 NP_061921.1 4 - 5 1902 76
XM_017004381.1 XP_016859870.1 3 - 4 1633 76
XM_017004382.1 XP_016859871.1 3 - 4 2418 76
GenBank mRNA AB075481.1 - 1 - 2 1351 76
AK000759.1 - 4 - 5 1881 76
AY116969.1 - 1 - 2 1468 76
BC001243.1 - 4 - 5 1807 76

View Full Product Details