The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035.2 | NP_001026.2 | 51 - 52 | 7949 | 131 |
XM_006711802.3 | XP_006711865.1 | 52 - 53 | 8173 | 131 | |
XM_006711803.3 | XP_006711866.1 | 52 - 53 | 8170 | 131 | |
XM_006711804.3 | XP_006711867.1 | 52 - 53 | 8173 | 131 | |
XM_006711805.3 | XP_006711868.1 | 51 - 52 | 8143 | 131 | |
XM_006711806.3 | XP_006711869.1 | 52 - 53 | 8173 | 131 | |
XM_006711807.3 | XP_006711870.1 | 52 - 53 | 8173 | 131 | |
XM_006711808.3 | XP_006711871.1 | 52 - 53 | 8173 | 131 | |
XM_006711809.3 | XP_006711872.1 | 52 - 53 | 8173 | 131 | |
XM_006711810.3 | XP_006711873.1 | 51 - 52 | 8140 | 131 | |
XM_017002028.1 | XP_016857517.1 | 51 - 52 | 8152 | 131 | |
GenBank mRNA | X98330.1 | - | 51 - 52 | 7950 | 131 |