The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035.2 | NP_001026.2 | 69 - 70 | 10264 | 80 |
XM_006711802.3 | XP_006711865.1 | 70 - 71 | 10488 | 80 | |
XM_006711803.3 | XP_006711866.1 | 70 - 71 | 10485 | 80 | |
XM_006711804.3 | XP_006711867.1 | 70 - 71 | 10488 | 80 | |
XM_006711805.3 | XP_006711868.1 | 69 - 70 | 10458 | 80 | |
XM_006711806.3 | XP_006711869.1 | 70 - 71 | 10488 | 80 | |
XM_006711807.3 | XP_006711870.1 | 70 - 71 | 10488 | 80 | |
XM_006711808.3 | XP_006711871.1 | 67 - 68 | 10251 | 80 | |
XM_006711810.3 | XP_006711873.1 | 69 - 70 | 10455 | 80 | |
XM_017002028.1 | XP_016857517.1 | 69 - 70 | 10467 | 80 | |
GenBank mRNA | AK294792.1 | - | 8 - 9 | 1282 | 80 |
X98330.1 | - | 69 - 70 | 10265 | 80 |