Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFN2
Entrez Gene ID
9927
Gene Name
mitofusin 2
Gene Aliases
CMT2A, CMT2A2, CPRP1, HMSN6A, HSG, MARF
Chromosome Location
Chr.1: 11980181 - 12013515 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127660.1 NP_001121132.1 3 - 4 624 68
NM_014874.3 NP_055689.1 4 - 5 769 68
XM_005263543.3 XP_005263600.1 5 - 6 782 68
XM_005263545.3 XP_005263602.1 4 - 5 632 68
XM_005263547.3 XP_005263604.1 4 - 5 664 68
XM_005263548.3 XP_005263605.1 5 - 6 799 68
GenBank mRNA AF036536.2 - 4 - 5 638 68
AK289828.1 - 3 - 4 569 68
AY028429.1 - 3 - 4 342 68
BC017061.1 - 4 - 5 499 68
BU178912.1 - 4 - 5 564 68
D86987.1 - 4 - 5 638 68

View Full Product Details