The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127660.1 | NP_001121132.1 | 4 - 5 | 780 | 83 |
NM_014874.3 | NP_055689.1 | 5 - 6 | 925 | 83 | |
XM_005263543.3 | XP_005263600.1 | 6 - 7 | 938 | 83 | |
XM_005263545.3 | XP_005263602.1 | 5 - 6 | 788 | 83 | |
XM_005263547.3 | XP_005263604.1 | 5 - 6 | 820 | 83 | |
XM_005263548.3 | XP_005263605.1 | 6 - 7 | 955 | 83 | |
GenBank mRNA | AF036536.2 | - | 5 - 6 | 794 | 83 |
AK289828.1 | - | 4 - 5 | 725 | 83 | |
AY028429.1 | - | 4 - 5 | 498 | 83 | |
BC017061.1 | - | 5 - 6 | 655 | 83 | |
D86987.1 | - | 5 - 6 | 794 | 83 |