The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080738.3 | NP_542776.1 | 5 - 6 | 451 | 238 |
NM_145861.2 | NP_665860.2 | 5 - 6 | 332 | 238 | |
GenBank mRNA | AK096339.1 | - | 5 - 6 | 451 | 238 |
AK290862.1 | - | 5 - 6 | 368 | 238 | |
AK314634.1 | - | 5 - 6 | 623 | 238 | |
AY028913.1 | - | 5 - 6 | 332 | 238 | |
AY028914.1 | - | 5 - 6 | 435 | 238 | |
AY071862.1 | - | 5 - 6 | 237 | 238 | |
BC114557.1 | - | 5 - 6 | 287 | 238 | |
BC114560.1 | - | 5 - 6 | 287 | 238 | |
BC128082.1 | - | 5 - 6 | 287 | 238 |