The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018841.5 | NP_061329.3 | 2 - 3 | 177 | 97 |
GenBank mRNA | AF119663.1 | - | 2 - 3 | 161 | 97 |
AF123766.1 | - | 2 - 3 | 168 | 97 | |
AF365871.1 | - | 2 - 3 | 104 | 97 | |
AK223000.1 | - | 2 - 3 | 119 | 97 | |
AK293101.1 | - | 2 - 4 | 97 | ||
AL832431.1 | - | 2 - 3 | 130 | 97 | |
BC005940.1 | - | 2 - 3 | 158 | 97 | |
BP236225.1 | - | 2 - 3 | 164 | 97 | |
BP236699.1 | - | 2 - 3 | 177 | 97 |