Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC16
Entrez Gene ID
23341
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Gene Aliases
-
Chromosome Location
Chr.1: 15526694 - 15571733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287811.1 NP_001274740.1 12 - 13 1762 96
NM_015291.3 NP_056106.1 13 - 14 1947 96
GenBank mRNA AB023179.2 - 13 - 14 1903 96
AK022530.1 - 4 - 5 314 96
AK292479.1 - 13 - 14 1879 96
BC031991.1 - 5 - 6 497 96
BC047363.1 - 13 - 14 1947 96
BX648982.1 - 13 - 14 1872 96
CR749259.1 - 12 - 13 1707 96
CR749567.1 - 12 - 13 1707 96

View Full Product Details