The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005575.2 | NP_005566.2 | 14 - 15 | 3067 | 93 |
NM_175920.3 | NP_787116.2 | 14 - 15 | 2859 | 93 | |
GenBank mRNA | AB208883.1 | - | 13 - 14 | 1941 | 93 |
AK314226.1 | - | 14 - 15 | 2609 | 93 | |
D50810.1 | - | 14 - 15 | 3024 | 93 | |
U62768.1 | - | 14 - 15 | 2614 | 93 | |
U62769.1 | - | 14 - 15 | 2869 | 93 |