The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005575.2 | NP_005566.2 | 16 - 17 | 3379 | 105 |
NM_175920.3 | NP_787116.2 | 16 - 17 | 3171 | 105 | |
GenBank mRNA | AB208883.1 | - | 15 - 16 | 2253 | 105 |
AK314226.1 | - | 16 - 17 | 2921 | 105 | |
U62768.1 | - | 16 - 17 | 2926 | 105 | |
U62769.1 | - | 16 - 17 | 3181 | 105 |